Please login to your online account to display your discounted pricing
Dynalon™ Straight-Sided Polypropylene Jars
Ideal for storing specimens, supplies or powders.
$26.95 - $60.40
Specifications
Closure Material Polypropylene
Autoclavable Yes
Closure Type Screw
Color Translucent
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Height (Metric) Price Quantity    
S63387
dynalon™
421135EMD
4 oz., 120mL 2.3 in. 60mm Pack of 12 Each for $40.25
Case of 3 Pack for $120.80
S63390
dynalon™
421165EMD
32 oz., 950mL 4 in. 100mm Pack of 6 Each for $60.40
Case of 4 Pack for $241.55
S63389
dynalon™
421155EMD
16 oz., 500mL 3.8 in. 97mm Pack of 6 Each for $41.00
Case of 4 Pack for $163.90
S63386
dynalon™
421125EMD
2 oz., 60mL 1.8 in. 47mm Pack of 12 Each for $26.95
Case of 4 Pack for $107.85
S63388
dynalon™
421145EMD
8 oz., 250mL 2.5 in. 66mm Pack of 6 Each for $29.50
Case of 6 Pack for $176.85
Description & Specifications
Specifications
Closure Material Polypropylene
Autoclavable Yes
Closure Type Screw
Color Translucent
Material Polypropylene
Includes White linerless screw closure
Max. Temperature (English) 275°F
Max. Temperature (Metric) 135°C
  • Translucent polypropylene is chemical-resistant
  • Autoclavable
  • Polypropylene screw caps
Polystyrene jars are also available (Cat. Nos. S63381 through -85).