Please login to your online account to display your discounted pricing

Stoddard Solvent, Fisher Chemical


A petroleum distillate used for dry cleaning

Quantity 200L

Manufacturer: fisher scientific company  BPS457200

 View more versions of this product

Catalog No. S457-200

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Boiling Point 149°C
Quantity 200L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless - Light Yellow
Vapor Pressure 0.37 - 0.39 kPa at 20°C
Vapor Density 4.8g/mol
ChemAlert Storage Symbol Red
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0
Acidity Pass Test
Chemical Name or Material Stoddard Solvent
Identification Pass Test
MDL Number MFCD00132767
CAS 64742-88-7
Boiling Range 90 to 100% less than 190°C

Specifications

Chemical Name or Material Stoddard Solvent
CAS 64742-88-7
MDL Number MFCD00132767
Accessories
 

Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0

WARNING!