Please login to your online account to display your discounted pricing

Stoddard Solvent, Fisher Chemical

A petroleum distillate used for dry cleaning

$402.23 - $402.23

Chemical Identifiers

Chemical Name or Material Stoddard Solvent
CAS 64742-88-7
MDL Number MFCD00132767
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S457-200

 
fisher scientific company
BPS457200
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S457-4

  Accessories
fisher scientific company
S4574
Glass Bottle 4L Each for $402.23
Case of 4 Each for $1,608.92
Description & Specifications

Specifications

Boiling Point 149°C
Quantity 200L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless - Light Yellow
Vapor Pressure 0.37 - 0.39 kPa at 20°C
Vapor Density 4.8g/mol
ChemAlert Storage Symbol Red
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0
Acidity Pass Test
Chemical Name or Material Stoddard Solvent
Identification Pass Test
MDL Number MFCD00132767
CAS 64742-88-7
Boiling Range 90 to 100% less than 190°C

Stoddard solvent, also known as mineral spirits, white spirit, petrolelum naptha, or paint thinner is a petroleum distillate used as a common solvent in paints, varnishes and asphalt, and in degreasing and dry cleaning. It is a mixture of aliphatic and alicyclic C7 to C12 hydrocarbons.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0

WARNING!