Stockwell Scientific Polystyrene Test Tubes
Precision-molded from premium-grade plastic and ideal for routine laboratory analysis and procedures
$255.75 - $255.75
Specifications
Color Clear
Material Polystyrene
Product Type Test Tube
Shape Round Bottom
Sterility Non-sterile
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Volume (Metric) Price Quantity  
22-046982 View Documents Stockwell Scientific
8560
5mL 12mm 75mm 5mL Case of 2000 Each for $255.75
Description
Description
  • Round bottom
  • Clear
  • Available in 5ml and 14ml capacities
  • 5mL tubes measure 12×75mm (0.47×2.95 in.)
  • 14mL tubes measure 16×100mm (0.63×3.94 in.)
Specifications
Specifications
Clear
Test Tube
Non-sterile
Polystyrene
Round Bottom