Please login to your online account to display your discounted pricing

Alfa Aesar™ STO-609, 98%

Chemical Identifiers

Chemical Name or Material STO-609
CAS 52029-86-4
Formula Weight 374.35
MDL Number MFCD06411456
Synonym 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67344LB0

 
alfa aesar™
J67344LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 374.35
Chemical Name or Material STO-609
Physical Form Solid
CAS 52029-86-4
Synonym 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate salt
Assay 98%
MDL Number MFCD06411456
Color Yellow
Quantity 5mg