Invitrogen™ STK4 (MST1) Recombinant Human Protein DFS Item
STK4 (MST1) is a serine/threonine kinase which may be involved in apoptosis.
Manufacturer:  Invitrogen™ LSPV4177
 View more versions of this product
Catalog No. PV4177
Description
Description
It is possible that the STK4 (MST1) protein induces the chromatin condensation observed in the apoptosis process.
Specifications
Specifications
NP_006273
Frozen
83.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK4 (MST1)
Baculovirus
Human
GST Tag
Documents