Learn More
Invitrogen™ STK39 (STLK3) Recombinant Human Protein
Recombinant full-length human STK39 (STLK3) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.
$800.00 - $12170.00
Specifications
For Use With (Application) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Formulation | 50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol |
Molecular Weight (g/mol) | 60.6kDa |
Storage Requirements | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Gene Alias | STK39, STLK3, DCHT, DKFZp686K05124, PASK, SPAK |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV6412
|
Invitrogen™
PV6412 |
10μg |
Each for $800.00
|
|
|||||
PV6413
|
Invitrogen™
PV6413 |
100μg |
Each for $2,715.00
|
|
|||||
PV6414
|
Invitrogen™
PV6414 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
STK39 (STLK3) is involved in the cellular stress response pathway. STK39 is activated in response to hypotonic stress leading to phosphorylation of several cation-chloride-coupled co-transporters. STK39 activates the p38 MAP kinase pathway and its interaction with p38 decreases during cellular stress. STK39 acts as an intermediate in the response to cellular stress. STK39 is also an independent risk factor for hypertension in men and its intragenic SNPs can interact and function in the control of blood pressure.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
60.6kDa | |
STK39, STLK3, DCHT, DKFZp686K05124, PASK, SPAK | |
Recombinant |
50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Human | |
His Tag |