missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ STK39 (STLK3) Recombinant Human Protein DFS Item

Recombinant full-length human STK39 (STLK3) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.

Manufacturer:  Invitrogen™ LSPV6414

 View more versions of this product

Catalog No. PV6414



Description

Description

STK39 (STLK3) is involved in the cellular stress response pathway. STK39 is activated in response to hypotonic stress leading to phosphorylation of several cation-chloride-coupled co-transporters. STK39 activates the p38 MAP kinase pathway and its interaction with p38 decreases during cellular stress. STK39 acts as an intermediate in the response to cellular stress. STK39 is also an independent risk factor for hypertension in men and its intragenic SNPs can interact and function in the control of blood pressure.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
60.6kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Yes
50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol
1mg
STK39, STLK3, DCHT, DKFZp686K05124, PASK, SPAK
Human
His Tag
Documents