STK33 Recombinant Human Protein DFS Item
STK33 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4343
 View more versions of this product
Catalog No. PV4343
Description
Description
STK33 is a serine⁄threonine kinase involved in signal transduction and cellular communication.
Specifications
Specifications
NP_112168
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
61.9kDa
70% by SDS-PAGE
–80°C
1 Each
STK33
STK33 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 500μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
CAMK Unique
12.5nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
His Tag
0.42mg/mL
65975
STK33 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
STK33 Recombinant Human Protein
Kinase assay
Baculovirus
STK33 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.