missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ STK32B (YANK2) Recombinant Human Protein DFS Item

STK32B or YANK2 is a serine/threonine protein kinase that contains motif for binding metal ions and nucleotides. Full-length recombinant human STK32B (YANK2) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

Manufacturer:  Invitrogen™ LSPV6407

 View more versions of this product

Catalog No. PV6407



Description

Description

The gene for STK32B is highly conserved among various species. STK32B has been associated with isolated cleft lip with or without cleft palate and cleft palate which are among the most common human birth defects of oral cleft cases. Long interspersed nuclear element-1 (LINE-1 or L1) mediated deletion of STK32B gene is observed in patients with Ellis-van Creveld syndrome with borderline intelligence.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
74.1kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
YANK2, STK32B, STK32, STKG6, HSA250839
Contains R198G SNP
Yes
Documents