Invitrogen™ STK25 (YSK1) Recombinant Human Protein DFS Item
STK25 (YSK1) is a serine/threonine kinase that belongs to the GCK family of kinases.
Manufacturer:  Invitrogen™ LSPV4065
 View more versions of this product
Catalog No. PV4065
Description
Description
STK25 (YSK1) has been implicated as an upstream regulator of MAP kinase signaling pathways.
Specifications
Specifications
NP_006365
Frozen
75.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK25 (YSK1)
Baculovirus
Human
GST Tag
Documents