missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

STK25 (YSK1) Recombinant Human Protein DFS Item

STK25 (YSK1) Recombinant Human Protein

$576.00 - $576.00

Specifications

Accession Number NP_006365
Assays STK25 (YSK1) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name STK25 (YSK1) Recombinant Human Protein
Concentration 0.37mg/mL
Expression System Baculovirus
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV3657 View Documents Life Technologies
LSPV3657
10μg Each for $576.00
 
Description

Description

STK25 (YSK1) is a serine⁄threonine kinase that belongs to the GCK family of kinases. It has been implicated as an upstream regulator of MAP kinase signaling pathways.
Specifications

Specifications

NP_006365
STK25 (YSK1) Recombinant Human Protein
Baculovirus
STE20
STK25 (YSK1) Recombinant Human Protein
STK25 (YSK1) Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
–80°C
Human
STK25 (YSK1) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
0.37mg/mL
STK25 (YSK1)
75.6kDa
1 Each
95% by SDS-PAGE
320nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL
Kinase assay
GST Tag

For Research Use Only. Not for use in diagnostic procedures.