Invitrogen™ STK23 (MSSK1) Recombinant Human Protein DFS Item
STK23 (MSSK1) is also known as SRPK3.
Manufacturer:  Invitrogen™ LSPV4198
 View more versions of this product
Catalog No. PV4198
Description
Description
STK23 (MSSK1) is a muscle-specific member of the SRPKs.
Specifications
Specifications
NP_055185
Frozen
86.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SRPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK23 (MSSK1)
Baculovirus
Human
GST Tag
Documents