Invitrogen™ STK22D (TSSK1) Recombinant Human Protein DFS Item
STK22D (TSSK1) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4063
 View more versions of this product
Catalog No. PV4063
Description
Description
STK22D (TSSK1) is essential to the function and production of sperm (spermiogenesis).
Specifications
Specifications
NP_114417
Frozen
46.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
TSSK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% NP40, 4mM DTT, 50% glycerol
1mg
RUO - research use only
STK22D (TSSK1)
Baculovirus
Human
His Tag
Documents