Invitrogen™ STK22B (TSSK2) Recombinant Human Protein DFS Item
STK22B (TSSK2) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4062
 View more versions of this product
Catalog No. PV4062
Description
Description
STK22B (TSSK2) is essential for the function and production of sperm (spermiogenesis). Deletions in the STK22B coding region correlate with numerous cardiovascular and glandular malformations.
Specifications
Specifications
NP_443732
Frozen
45.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
G11 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK22B (TSSK2)
Baculovirus
Human
His Tag
Documents