Invitrogen™ STK17A (DRAK1) Recombinant Human Protein DFS Item
STK17A (DRAK1) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4061
 View more versions of this product
Catalog No. PV4061
Description
Description
STK17A (DRAK1) is localized in the nucleus and is involved in apoptosis.
Specifications
Specifications
NP_004751
Frozen
74kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
DAPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
STK17A (DRAK1)
Baculovirus
Human
GST Tag
Documents