Stir-Pak™ General-Purpose Mixers
High-quality, reliable mixers featuring full-range performance.
Manufacturer:  Stir-Pak™ 0455500
Catalog No. 13-200-284
Includes:
  • 3/8 (9.5mm) dia x 12 in. (305mm) L, 316 stainless steel shaft
  • 1.5 in. (38mm) dia. Propeller
  • 6 in. (152mm) mounting rod
  • 6 ft. (1.8m) grounded cord with power supply
Description
Description
Ideal for routine mixing of water-like viscous liquids.
  • Brushed DC motor has a compact speed controller integrated by a custom anodized aluminum potentiometer
  • LED light indicates situations when the motor is turned on/overloaded/overtemperature
  • Keyless chuck accepts mixing shafts from 5/16 to 3/8 in. (8 to 10mm) dia.
  • Gives full-range performance
Warranty
Two year
Specifications
Specifications
Each
Ideal for routine, light duty mixing of water like viscosities
40 to 4000rpm
115/230VAC 50Hz
50Hz
115/230VAC
0.37 in. dia. x 12 in. L, 316 stainless steel shaft, 1.5 in. dia. propeller, 6 in. mounting rod, 72 in. grounded cord with power supply
Documents