Please login to your online account to display your discounted pricing

STF 31, Tocris™

GLUT1 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material STF 31
Assay Percent Range >99%
Molecular Formula C23H25N3O3S
Formula Weight 423.53
Synonym 4-[[[[4-(1, 1-Dimethylethyl)phenyl]sulfonyl]amino]methyl]-N-3-pyridinylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

448410

 
r & d systems
RDS448410
10mg Each for $252.39

448450

 
r & d systems
RDS448450
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 423.53
Quantity 10mg
Chemical Name or Material STF 31
Synonym 4-[[[[4-(1, 1-Dimethylethyl)phenyl]sulfonyl]amino]methyl]-N-3-pyridinylbenzamide
Molecular Formula C23H25N3O3S
CAS 724741-75-7
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature