Please login to your online account to display your discounted pricing

STF 083010, Tocris™

Inhibits IRE1α endonuclease activity

$225.77 - $914.83

Chemical Identifiers

Chemical Name or Material STF 083010
Assay Percent Range >98%
Molecular Formula C15H11NO3S2
Formula Weight 317.38
Synonym N-[(2-Hydroxy-1-naphthalenyl)methylene]-2-thiophenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

450910

 
r & d systems
RDS450910
10mg Each for $225.77

450950

 
r & d systems
RDS450950
50mg Each for $914.83
Description & Specifications

Specifications

Formula Weight 317.38
Quantity 10mg
Chemical Name or Material STF 083010
Assay Percent Range >98%
Molecular Formula C15H11NO3S2
CAS 307543-71-1
Recommended Storage Store at -20°C
Synonym N-[(2-Hydroxy-1-naphthalenyl)methylene]-2-thiophenesulfonamide
Safety and Handling

Store at -20°C