missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Reinforced Membrane Filters

Contain no asbestos or fiberglass; made from non-shedding materials.

$107.00 - $229.00

Specifications

Autoclavable Autoclavable
Color White
Filter Type Screen filter
Material Polymers of Cellulose Ester
Pore Size 0.5 μm
View More Specs

Products 7
Catalog Number Mfr. No. Diameter (Metric) Disc Flow Rate Grade Particle Retention Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Disc Flow Rate Grade Particle Retention Price Quantity  
RW0309000
Accessories View Documents Promotion Details
MilliporeSigma™
RW0309000
9 cm 50 mL/min/ cm2 RW03 0.45 µm
Pack of 100 Each for $201.00
 
RW0609000
Accessories View Documents Promotion Details
MilliporeSigma™
RW0609000
9 cm 150 mL/min/ cm2 RW06 0.8 µm
Pack of 100 Each for $201.00
 
RW1904700
Accessories View Documents Promotion Details
MilliporeSigma™
RW1904700
4.7 cm 260 mL/min/ cm2 RW19 1.2 µm
Pack of 100 Each for $107.00
 
RW1914250
Accessories View Documents Promotion Details
MilliporeSigma™
RW1914250
14.2 cm 260 mL/min/ cm2 RW19 1.2 µm
Pack of 50 Each for $229.00
 
RW0304700
Accessories View Documents Promotion Details
MilliporeSigma™
RW0304700
4.7 cm 50 mL/min/ cm2 RW03 0.45 µm
Pack of 100 Each for $107.00
 
RW0604700
Accessories View Documents Promotion Details
MilliporeSigma™
RW0604700
4.7 cm 150 mL/min/ cm2 RW06 0.8 µm
Pack of 100 Each for $107.00
 
RW0614250
Accessories View Documents Promotion Details
MilliporeSigma™
RW0614250
14.2 cm 150 mL/min/ cm2 RW06 0.8 µm
Pack of 50 Each for $229.00
 
Description

Description

Reinforced prefilter membranes can be used to remove contaminants from a variety of liquids and gases. The filters have a high dirt-holding capacity and a low pressure drop, which makes them ideally suited for reducing contaminant levels in advance of sterilizing-grade filters. Unlike traditional prefilter materials that contain asbestos or fiberglass, our reinforced prefilter membranes are made from non-shedding materials. These filters retain contaminants on the surface of the cellulose membrane.

Features & Benefits:
  • Prefilter with high dirt-loading capacity for greater throughput
  • Non-shedding substrate will not unload fibers downstream

Applications: Prefiltration Ahead of Sterilizing-Grade Filters

Specifications

Specifications

Autoclavable
Screen filter
0.5 μm
Hydrophilic
158°F
0.007 in.
Filter Membranes
White
Polymers of Cellulose Ester
Sterile
Plain
70°C
200 µm
Hydrophilic
SDS
missing translation for 'documents'

missing translation for 'documents'