Learn More
BIOTIX™ Sterile uTIP Filter Tip Reload Universal Compatible Pipette Tips
BIOTIX™ Sterile uTIP™ Filter Tip Reload Universal Pipette Tips Sustainable Packaging, with Flexfit™ to lower insertion and ejection force by approx. 70%. Manufactured with pure resin that is virgin polypropylene. RNase, DNase, Pyrogen, and PCR Inhibitor free.
$301.97 - $377.43
Specifications
| Filtered | Filtered |
|---|---|
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Clean Claims | Fully Recyclable |
| Catalog Number | Mfr. No. | Volume (Metric) | Length (Metric) | Color Code | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Length (Metric) | Color Code | Price | Quantity | |||
|
01-671-702
|
BIOTIX™
63305488 |
1200 μL | 98.30 mm | Blue |
Pack of 960 Each for $377.43
Case of 4 Pack for $1,509.72
|
|
|||
|
01-671-703
|
BIOTIX™
63305487 |
2 μL | 31.24 mm | Red |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-707
|
BIOTIX™
63301928 |
1000 μL | 81.79 mm | Blue |
Pack of 960 Each for $377.43
Case of 4 Pack for $1,509.72
|
|
|||
|
01-671-709
|
BIOTIX™
63301926 |
200 μL | 59.54 mm | Yellow |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-710
|
BIOTIX™
63301925 |
100 μL | 59.54 mm | Yellow |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-711
|
BIOTIX™
63301924 |
20 μL | 50.75 mm | Yellow |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-712
|
BIOTIX™
63301923 |
10 μL | 45.84 mm | Red |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-713
|
BIOTIX™
63301922 |
10 μL | 31.24 mm | Red |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
|
01-671-708
|
BIOTIX™
63301927 |
300 μL | 59.54 mm | Green |
Pack of 960 Each for $301.97
Case of 5 Pack for $1,509.85
|
|
|||
Description
BIOTIX™ Sterile uTIP™ Filter Tip Reloads, are manufactured in sustainable benchtop and user friendly packaging. Reduce plastic waste by 30% and easily refill your racks with utip Filter Tip Reloads.
- Fits universal pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
- Pure resin made of virgin polypropylene
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Specifications
| Filtered | |
| Clear | |
| Fully Recyclable | |
| Pipette Tips | |
| Universal Fit | |
| 100% Virgin Polypropylene | |
| Certified Quality |
| Sterile | |
| Not Autoclavable | |
| Blister Refill | |
| Single-use | |
| Yes | |
| Conical |