Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
$406.79 - $406.79
Specifications
For Use With (Equipment) Genesis, Freedom Evo, MiniPrep and Carvro
Sterility Sterile
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Filtered Packaging Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Filtered Packaging Volume (Metric) Price Quantity  
10-700-100 Thermo Scientific
LMF17596RS1000
Clear Yes 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case 1000μL Pack of 960 Each for $406.79
Case of 5 Pack for $2,033.95
Specifications
Description
Increase pipetting performance with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
Warranty: A five (5)-year shelf life from the date of manufacture is recommended.
Description & Specifications
Specifications
Genesis, Freedom Evo, MiniPrep and Carvro
Sterile