Please login to your online account to display your discounted pricing

Thermo Scientific Sterile Automation Tips for Tecan™ Liquid Handling Platforms


Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.

Manufacturer: thermo scientific  LMF17596RS1000

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 10-700-100

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

For Use With (Equipment) Genesis, Freedom Evo, MiniPrep and Carvro
Volume (Metric) 1000μL
Product Line Tecan
Sterility Sterile
Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Filtered Yes
Color Clear
Sterile Yes
Features Filtered
Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Automated Workstation Brand Tecan
Automated Workstation Model Genesis, Freedom Evo, MiniPrep and Carvro
Volume (Metric) 1000µL

Increase pipetting performance with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)