missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Propper Steri-Wrap™ III Autoclavable Wraps
Ideal for a variety of uses including OR and CSR wrapping
Manufacturer: Propper 31305400
Description
- Excellent microbial barrier—virtually lint-free and completely liquid repellent
- Particularly recommended for applications where strength is required for heavy items such as instrument trays and basin sets
- Made from single-use, nonwoven, and tightly anchored fabric
- Can be used on either side without risk of delaminating
Order Info
Order autoclavable tape separately
Specifications
Yes | |
OR and CSR wrapping | |
137.1cm | |
Non-woven Fabric | |
137.1cm |
54 x 54 in. (137.1 x 137.1cm) | |
54 in. | |
Steri-Wrap III | |
54 in. |