Please login to your online account to display your discounted pricing
Fisher Scientific™ Stereomaster™ Microscope Eyepieces with Micrometer Reticle
For Fisher Scientific Stereomaster microscopes
Products
Catalog Number Mfr. No. Price Quantity    
12-562-ER6
fisher scientific™
MPEP10XRG
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
  • Reticle grid is 10 × 10mm
  • Grid has 1mm divisions