Please login to your online account to display your discounted pricing

Step-On™ Waste Containers

Quiet, heavy-duty pedal for hands-free sanitary use

$599.55 - $599.55

Specifications

Certifications/Compliance FM, OSHA 29 CFR 1910.1030
Color Red
Product Type Waste Container
For Use With (Application) Ideal for use in patient rooms, lab procedure rooms and emergency rooms
Includes Slow close integrated lid
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Height (English) Material Price Quantity    

S94850

 
fisher science education
6147-00/E
30 gal. 34.25 in. Stainless Steel Each for $599.55
Description & Specifications

Specifications

Certifications/Compliance FM, OSHA 29 CFR 1910.1030
Color Red
Product Type Waste Container
For Use With (Application) Ideal for use in patient rooms, lab procedure rooms and emergency rooms
Includes Slow close integrated lid
Shape Oval

  • Self-closing lid with an integrated lid damper that reduces noise and keeps odors and bacteria from escaping container
  • Ideal for use in patient rooms, lab procedure rooms and emergency rooms
  • Available in impact-resistant plastic or stainless-steel/plastic hybrid construction
  • 23 or 30 gal. capacities

FM approved.

Assists in complying with OSHA 29 CFR 1910.1030 Occupational Exposure to Bloodborne Pathogens