Fisherbrand™ Steel Tall Cabinets
Fisherbrand™ Steel Tall Cabinets are free standing 2 door cabinets and come with locks.
Manufacturer:  Fisherbrand™ FJTP30348L
 View more versions of this product
Catalog No. 06000047
Description
Description
  • Standard with locks
  • Five shelves are included and are adjuatble in half-inch increments
  • Must be anchored to wall
  • When installing tall cases under soffits, 85-1/2 in. minimum height is required from finished floor to finished soffit
Compliance
SEFA 8
Specifications
Specifications
Solid Door Tall Cabinet
White
55.9cm
214.9cm
Steel
Tall Cabinet
121.9cm
SEFA 8
22 in.
84.625 in.
Five adjustable shelves, locking doors
2
48 in.
Documents