Please login to your online account to display your discounted pricing

Steel


Quantity 500g
Packaging Steel Wool

Manufacturer: fisher science education  S25586

 View more versions of this product

Catalog No. S25586

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Color Gray
Chemical Name or Material Steel
Shelf Life 5 years
Packaging Steel Wool
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Steel
Accessories
 

5 years

SDS & Safety Documents