missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stearyldiethanolamine, TCI America™

$285.70 - $936.55

Chemical Identifiers

CAS 10213-78-2
Molecular Formula C22H47NO2
Molecular Weight (g/mol) 357.623
MDL Number MFCD00059003
InChI Key NDLNTMNRNCENRZ-UHFFFAOYSA-N
Synonym 2,2′-(Octadecylimino)diethanol
PubChem CID 61494
IUPAC Name 2-[2-hydroxyethyl(octadecyl)amino]ethanol
SMILES CCCCCCCCCCCCCCCCCCN(CCO)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O001725G View Documents TCI America
O001725G
25g Each for $936.55
 
O00175G View Documents TCI America
O00175G
5g Each for $285.70
 
Specifications

Chemical Identifiers

10213-78-2
357.623
NDLNTMNRNCENRZ-UHFFFAOYSA-N
61494
CCCCCCCCCCCCCCCCCCN(CCO)CCO
C22H47NO2
MFCD00059003
2,2′-(Octadecylimino)diethanol
2-[2-hydroxyethyl(octadecyl)amino]ethanol

Specifications

White
25g
C22H47NO2
2,2′-(Octadecylimino)diethanol
CCCCCCCCCCCCCCCCCCN(CCO)CCO
357.623
357.62
≥98.0% (T)
51°C
10213-78-2
MFCD00059003
NDLNTMNRNCENRZ-UHFFFAOYSA-N
2-[2-hydroxyethyl(octadecyl)amino]ethanol
61494
Crystal-Powder at 20°C
Stearyldiethanolamine
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0353&(7)-0060

TSCA : Yes