missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stearylamine 85.0+%, TCI America™

Manufacturer:  TCI America O0014500G

 View more versions of this product

Catalog No. O0014500G



Specifications

Specifications

Stearylamine
White
500g
C18H39N
3077
REYJJPSVUYRZGE-UHFFFAOYSA-N
octadecan-1-amine
15793
269.52
≥85.0% (GC)
349°C
53°C
124-30-1
MFCD00008159
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
CCCCCCCCCCCCCCCCCCN
269.517
CHEBI:63866
Crystal-Powder at 20°C

Chemical Identifiers

124-30-1
269.517
REYJJPSVUYRZGE-UHFFFAOYSA-N
15793
octadecan-1-amine
C18H39N
MFCD00008159
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
CHEBI:63866
CCCCCCCCCCCCCCCCCCN
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0133&(2)-0176&(2)-0185

RTECSNumber : RG4150000

TSCA : Yes

Documents