Please login to your online account to display your discounted pricing

Stearyl stearate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368110050

 View more versions of this product

Catalog No. AC368110050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder or Platelets
Formula Weight 536.97
Quantity 5g
Color White
Chemical Name or Material Stearyl stearate
Assay Percent Range 98%
Linear Formula CH3(CH2)16COO(CH2)17CH3
CAS 2778-96-3
Packaging Glass bottle
Name Note 98%
Molecular Formula C36H72O2
MDL Number MFCD00056225
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Stearyl stearate
Name Note 98%
CAS 2778-96-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C36H72O2
Linear Formula CH3(CH2)16COO(CH2)17CH3
Formula Weight 536.97
MDL Number MFCD00056225