missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Stearyl stearate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 368110010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Stearyl stearate | |
2778-96-3 | |
97.5% min. (GC) | |
C36H72O2 | |
MFCD00056225 | |
stearyl stearate, octadecyl stearate, octadecanoic acid, octadecyl ester, stearic acid stearyl ester, unii-5wx2egd0dk, stearic acid, stearyl ester, octadecanyl octadecanoate, 5wx2egd0dk, stearic acid, octadecyl ester, cyclochem ss | |
CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC | |
536.97 | |
536.97 |
98% | |
Authentic | |
Glass bottle | |
CH3(CH2)16COO(CH2)17CH3 | |
1g | |
NKBWPOSQERPBFI-UHFFFAOYSA-N | |
octadecyl octadecanoate | |
17720 | |
98% |
Chemical Identifiers
2778-96-3 | |
536.97 | |
NKBWPOSQERPBFI-UHFFFAOYSA-N | |
17720 | |
CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC |
C36H72O2 | |
MFCD00056225 | |
stearyl stearate, octadecyl stearate, octadecanoic acid, octadecyl ester, stearic acid stearyl ester, unii-5wx2egd0dk, stearic acid, stearyl ester, octadecanyl octadecanoate, 5wx2egd0dk, stearic acid, octadecyl ester, cyclochem ss | |
octadecyl octadecanoate |
Safety and Handling
EINECSNumber : 220-476-5
RUO â Research Use Only