missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stearyl Glycyrrhetinate 98.0+%, TCI America™

Manufacturer:  TCI America S09885G

 View more versions of this product

Catalog No. S09885G



Specifications

Specifications

Stearyl Glycyrrhetinate
77°C
13832-70-7
MFCD00674112
WNIFXKPDILJURQ-OVDQTPITSA-N
octadecyl (2S,4aS,6aR,6bR,10S,12aS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
122130446
Crystal-Powder at 20°C
White-Yellow
5g
C48H82O4
Glycyrrhetinic Acid Stearyl Ester, Octadecyl Glycyrrhetinate, Glycyrrhetinic Acid Octadecyl Ester
CCCCCCCCCCCCCCCCCCOC(=O)C1(CCC2(CCC3(C(=CC(=O)C4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C)C
723.18
723.18
≥98.0% (HPLC)

Chemical Identifiers

13832-70-7
723.18
WNIFXKPDILJURQ-OVDQTPITSA-N
122130446
CCCCCCCCCCCCCCCCCCOC(=O)C1(CCC2(CCC3(C(=CC(=O)C4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C)C
C48H82O4
MFCD00674112
Glycyrrhetinic Acid Stearyl Ester, Octadecyl Glycyrrhetinate, Glycyrrhetinic Acid Octadecyl Ester
octadecyl (2S,4aS,6aR,6bR,10S,12aS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0722

TSCA : No

Documents