missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Stearic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1224422

 View more versions of this product

Catalog No. AAA1224422



Description

Description

A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A saturated fatty acid Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

Solubility
Soluble in ethanol, dimethyl sulfoxide, dimethyl formamide, carbon terachloride, toluene, chloroform, acetone and carbondisulfide. Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Specifications

Stearic Acid
69°C to 72°C
232°C (15 mmHg)
98%
MFCD00002752
608585
Soluble in ethanol,dimethyl sulfoxide,dimethyl formamide,carbon terachloride,toluene,chloroform,acetone and carbondisulfide. Insoluble in water.
CCCCCCCCCCCCCCCCCC(O)=O
284.48
CHEBI:28842
57-11-4
0.941
196°C (384°F)
C18H36O2
100 g
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
QIQXTHQIDYTFRH-UHFFFAOYSA-N
octadecanoic acid
5281
284.48

Chemical Identifiers

57-11-4
284.48
QIQXTHQIDYTFRH-UHFFFAOYSA-N
5281
octadecanoic acid
C18H36O2
MFCD00002752
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
CHEBI:28842
CCCCCCCCCCCCCCCCCC(O)=O
Safety and Handling

Safety and Handling

H500

EINECSNumber : 200-313-4

RTECSNumber : WI2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only