missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ STE buffer soln.

Specifications

Chemical Name or Material STE buffer soln.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60265K2 View Documents Alfa Aesar™
J60265K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ60265K3 View Documents Alfa Aesar™
J60265K3
2L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
STE buffer is a DNA resuspension and storage buffer used in plasmid Kits for plasmid purification and in some plasmid supplementary protocols. Used in DNA extraction, DNA electrophoresis and many other lab protocols.

Solubility
Soluble in water.

Notes
Contains 100mM NaCl, 10mM Tris-HCl, 1mM EDTA, pH 8.0.
Specifications

Specifications

STE buffer soln.