Please login to your online account to display your discounted pricing

Stattic, Tocris

Selective STAT3 inhibitor

$95.70 - $392.76

Chemical Identifiers

Chemical Name or Material Stattic
Assay Percent Range >99%
Molecular Formula C8H5NO4S
Formula Weight 211.19
Synonym 6-Nitrobenzo[b]thiophene 1, 1-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

279850

 
r & d systems
RDS279850
50mg Each for $392.76

279810

 
r & d systems
RDS279810
10mg Each for $95.70
Description & Specifications

Specifications

Formula Weight 211.19
Quantity 50mg
Assay Percent Range >99%
CAS 19983-44-9
Chemical Name or Material Stattic
Synonym 6-Nitrobenzo[b]thiophene 1, 1-dioxide
Molecular Formula C8H5NO4S
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C