StatSpin™ CytoFuge™ 2 Personal Cytocentrifuge System Accessories: Gaskets and Concentrators
For use with CytoFuge 2 Personal Cytocentrifuge System
$46.39 - $226.51
Specifications
For Use With (Equipment) CytoFuge 2
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
22-020738 Beckman Coulter Canada Lp
CC01
3 well Cell Concentrators, Disposable 10/Pk. Pack of 10 Each for $46.39
22-020737 Beckman Coulter Canada Lp
CC03
1 well Cell Concentrators, Disposable 10/Pk. Pack of 10 Each for $60.31
22-029484 Beckman Coulter Canada Lp
GK03
Silicone Gasket for 3 well cell Concentrator 100/Pk. Pack of 100 Each for $226.51
Specifications
Description
Handle all specimens from routine body fluid preparations to multiple cell concentrations or controls on the same slide.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
CytoFuge 2