missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Statsafe™ 6000

Chemical Identifiers

UN Number UN1993
Chemical Name or Material Statsafe(TM) 6000
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5578722 SDS Alfa Aesar™
H5578722
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5578714 SDS Alfa Aesar™
H5578714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5578736 SDS Alfa Aesar™
H5578736
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Static dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. The Protein Crystallography Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).

Solubility
Not miscible with water.

Notes
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.
Description & Specifications

Chemical Identifiers

UN1993
Statsafe(TM) 6000

Specifications

0.805g/mL
UN1993
Pale yellow
100g
Liquid
94°C
<−5°C (−23°F)
Statsafe(TM) 6000

Safety and Handling

GHS H Statement
H225-H304-H314-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA