missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Statsafe^t 6000, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5578714
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsStatic dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. The Protein Crystallography Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).
Solubility
Not miscible with water.
Notes
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.
Specifications
Statsafe™ 6000 | |
UN1993 | |
Not miscible with water. |
0.805 | |
25g | |
<−5°C (−23°F) |
RUO â Research Use Only