missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Statsafe™ 6000

Manufacturer:  Alfa Aesar™ H5578736

 View more versions of this product

Catalog No. AAH5578736

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
Static dissipater used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss. The Protein Crystallography Extension Kit substantially increases the screening range of the Basic Crystallography Kit by providing a broader range of organic solvents, PEGs, and additives (divalent ions and stabilizers).

Solubility
Not miscible with water.

Notes
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.
Specifications

Specifications

Statsafe™ 6000
Liquid
<−5°C (−23°F)
UN1993
94°C
500g
Documents