missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starplex Scientific Histoplex™ Tissue Cassettes with Micromesh Chamber

Innovative micromesh chamber securely retains fragments, leaving ample room for additional larger specimens

$553.59 - $553.59

Specifications

Closure Type Clam-Shell
Description Tissue/Biopsy Cassette
Material Acetal Polymer
Pore Style Slots
Writing Face Angle 45°
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22-146-427 Fisher Scientific
TCRED
Red Case of 1500 Each for $553.59
Specifications

Description

  • Typical-looking tissue cassette designed with a corner section for use as a biopsy cassette
  • Fine micromesh chamber (0.23mm) is ideal for tissue fragments
  • Effective for embedding, processing, and storing standard tissue specimens and fragments in the same cassette
  • Durable acetyl polymer resists the chemical action of histological agents
  • Flow-through slots maximize fluid exchange
  • Textured 45° writing angle, suitable for labeling devices
  • Clam-shell design snaps closed and reopens with ease
  • Reduce inventory without giving up quality
  • Available in an assortment of non-metallic colors
  • Conveniently packaged 500 cassettes per dispenser box, three boxes per case

Description & Specifications

Specifications

Clam-Shell
Acetal Polymer
45°
Tissue/Biopsy Cassette
Slots