Starplex™ Scientific Histoplex™ Histology Containers
Specifically engineered for the storage of histology specimens
Manufacturer:  Starplex™ Scientific H50002
 View more versions of this product
Catalog No. 15-182-78
Description
Description
Specifically engineered for the storage of histology specimens
  • O-ring polyethylene screw cap forms a seal that reduces most formalin leakage and evaporation
  • Durable polypropylene containers are translucent, flexible, resist cracking and splitting, and eliminate problems associated with formalin evaporation
  • 40mL to 1L containers have graduations for easy fill line viewing
  • 2 to 8L containers are white polypropylene with unique tamper-evident snap-on lids
Specifications
Specifications
Non-sterile
Polypropylene
5L
In case
White
Snap-on
White
Yes
Documents