Please login to your online account to display your discounted pricing

Starch, soluble, ACS (for iodometry), Alfa Aesar™

$31.34 - $31.34

Chemical Identifiers

Chemical Name or Material Starch
CAS 9005-84-9
Name Note soluble
Purity Grade ACS Grade
Purity Grade Notes For iodometry
MDL Number MFCD00082026
Merck Index 14,8799
Synonym Amylodextrin
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3670318

  SDS
alfa aesar
3670318
Each for $31.34
Description & Specifications

Specifications

Melting Point 256° to 258°C (dec.)
Purity Grade ACS Grade
Quantity 50g
Purity Grade Notes For iodometry
CAS 9005-84-9
Solubility Information Soluble in hot water
TSCA TSCA
Merck Index 14,8799
Chemical Name or Material Starch
Synonym Amylodextrin
Name Note soluble
Odor Odorless
Physical Form Powder
MDL Number MFCD00082026