missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Starch, modified, insolubles 0.01% max, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03667336
Description
Starch, modified is used in the production of facial cleanser, oil controller, sunscreen cream, moisturizer, toners/astringents, facial moisturizer/treatment and anti-aging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Starch, modified | |
Powder | |
C12H22O11 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
500 g | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:18167 |
Insolubles 0.01% max | |
9005-84-9 | |
MFCD00082026 | |
Instantly soluble in water | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
439341 | |
Odorless |
Chemical Identifiers
9005-84-9 | |
342.297 | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Safety and Handling
EINECSNumber : 232-686-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only