missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch-Iodide Solution, Mercury Free, Certified, LabChem™

$44.06 - $44.06

Chemical Identifiers

CAS 7732-18-5,9005-25-8,69-72-7,7681-11-0
CAS Max % 99.32,0.5,0.13,0.05
Linear Formula KI
Formula Weight 166
Chemical Name or Material Starch-Iodide Solution, Mercury Free
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC253307 Lab Chem
LC253307
125mL Each for $44.06
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,9005-25-8,69-72-7,7681-11-0
KI
Starch-Iodide Solution, Mercury Free
99.32,0.5,0.13,0.05
166
Certified Grade

Specifications

7732-18-5,9005-25-8,69-72-7,7681-11-0
Liquid
166
Passes Test
Soluble in water
Passes Test
Fume; Carbon monoxide; Carbon dioxide
125mL
Certified Grade
99.32,0.5,0.13,0.05
KI
1g/mL
Poly Bottle
1g/mL
Room Temperature
Slightly turbid
Starch-Iodide Solution, Mercury Free

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.