missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations

For iodometric titrations

Manufacturer:  Alfa Aesar™ 47209AP

 View more versions of this product

Catalog No. AA47209AP



Specifications

Specifications

Starch indicator solution 1%
9005-84-9
MFCD00082026
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
342.297
CHEBI:18167
500mL
w/v aqueous solution for iodometric titrations
C12H22O11
14,8799
GUBGYTABKSRVRQ-ASMJPISFSA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
439341
Liquid

Chemical Identifiers

9005-84-9
342.297
GUBGYTABKSRVRQ-ASMJPISFSA-N
439341
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C12H22O11
MFCD00082026
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
CHEBI:18167
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
Safety and Handling

Safety and Handling

RTECSNumber : GM5090000

TSCA : TSCA

SDS Documents