Please login to your online account to display your discounted pricing

Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1L, Alfa Aesar™

For Iodometric Titrations

$54.69 - $54.69

Chemical Identifiers

Chemical Name or Material Starch indicator solution
CAS 9005-84-9
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 100%
MDL Number MFCD00082026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88632LQ

  SDS
alfa aesar
88632LQ
1 unit Each for $54.69

AA88632LS

  SDS
alfa aesar
88632LS
6 units CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 unit
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Concentration or Composition (by Analyte or Components) Starch: 20%; Water: 80%
MDL Number MFCD00082026
CAS 9005-84-9
Assay Percent Range 100%
CAS 7732-18-5
Chemical Name or Material Starch indicator solution