missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 1%, LabChem™

Manufacturer:  LabChem LC253105

 View more versions of this product

Catalog No. LC253105



Specifications

Specifications

Starch Indicator
98.9,1,0.1
Passes Test
1%
Cubitainer™
1g/mL
Fume; Carbon monoxide; Carbon dioxide
Colorless to Pink
Preserved with Salicylic Acid (Mercury Free)
9005-25-8,69-72-7
Liquid
Certified
Soluble in water
Passes Test
1g/mL
20L
Documents
Promotions

Promotions