missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.5%, LabChem™

Manufacturer:  LabChem LC253001

 View more versions of this product

Catalog No. LC253001



Specifications

Specifications

Starch Indicator
99.4,0.5,0.1
Passes Test
0.5%
Poly Bottle
1g/mL
Fume; Carbon monoxide; Carbon dioxide
Undesignated
Preserved with Salicylic Acid (Mercury Free)
9005-25-8,69-72-7
Liquid
Certified
Soluble in water
Passes Test
1g/mL
500mL
Documents
Promotions

Promotions