Please login to your online account to display your discounted pricing

Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 0.2%, LabChem™

$22.00 - $22.00

Chemical Identifiers

Chemical Name or Material Starch Indicator
CAS 7732-18-5
CAS Max % 99.7
CAS 9005-25-8
CAS Max % 0.2
CAS 69-72-7
CAS Max % 0.1
Name Note Preserved with Salicylic Acid (Mercury Free)
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC252907

 
lab chem
LC252907
Poly Bottle 125mL Each for $22.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 125mL
Recommended Storage Room Temperature
Name Note Preserved with Salicylic Acid (Mercury Free)
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
CAS Max % 99.7
Solubility Soluble in water
Suitability Passes Test
Specific Gravity 1g/mL
Identification Passes Test
Assay 0.2%
CAS Max % 0.2
CAS 69-72-7
Chemical Name or Material Starch Indicator
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Packaging Poly Bottle
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS 9005-25-8
CAS Max % 0.1
Physical Form Liquid
CAS 7732-18-5
Density 1g/mL
Color Slightly turbid
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature